Learn More
Medchemexpress LLC N4-acetylsulfamethazine | 100-90-3 | >95.0% | 320.37 g/mol | C14H16N4O3S | 5 MG
SDP
Catalog No. 5000282053
Encompass_Preferred
N4-Acetylsulfamethazine is the N4-acetylated metabolite of sulfamethazine supplied for research use as a reference standard and reagent for metabolic, residue, and analytical studies. The material is an off-white to light-yellow solid and should be stored at low temperatures to maintain stability.
  • Serves as a metabolite reference for sulfamethazine in metabolic and residue studies.
  • Suitable as an analytical standard for HPLC and mass spectrometry assays.
  • Off-white to light-yellow solid for easy visual identification.
  • Powder stable at -20°C for long-term storage.
  • Available in small sample quantities for method development and validation.
Catalog No. 50-002-82053 Supplier Medchemexpress LLC Supplier No. HYW2908395MG
N4-Acetylsulfamethazine is the N4-acetylated metabolite of sulfamethazine supplied for research use as a reference standard and reagent for metabolic, residue, and analytical studies. The material is an off-white to light-yellow solid and should be stored at low temperatures to maintain stability.
  • Serves as a metabolite reference for sulfamethazine in metabolic and residue studies.
  • Suitable as an analytical standard for HPLC and mass spectrometry assays.
  • Off-white to light-yellow solid for easy visual identification.
  • Powder stable at -20°C for long-term storage.
  • Available in small sample quantities for method development and validation.