Learn More
AdipoGen N4-Acetylsulfamethoxazole
SDP
Supplier:  AdipoGen CDXA0291M050
Encompass_Preferred
Chemical. CAS 21312-10-7. Formula C12H13N3O4S. MW 295.3. Synthetic. This is a metabolite of the sulfonamide bacteriostatic antibiotic sulfamethoxazole. Sulfamethoxazole is metabolized via acetylation, catalyzed by liver extramitochondrial N-acetyl transferases. Acetyl-Sulfamethoxazole is excreted in urine. Both compounds can be used as a probe for the molecular percentage enrichement of liver extramitochondrial acetyl-CoA. Acetyl-Sulfamethoxazole can also be used as a reference for sulfamethoxazole impurity and waste determination.
Catalog No. 50-256-1765