Learn More
Cayman Chemical N4-benzoyl-2'-Deoxycytidine, Purity: ≥98%, CAS: 4836-13-9, 50 g
SDP
Catalog No. 502942038
Encompass_Preferred
A building block; has been used in the synthesis of nucleosides and nucleotides with anticancer activity, as well as oligonucleotides
Catalog No. 50-294-2038 Supplier Cayman Chemical Supplier No. 3993450 G
A building block; has been used in the synthesis of nucleosides and nucleotides with anticancer activity, as well as oligonucleotides