Learn More
Cayman Chemical N4-benzoyl-2'-Deoxycytidine, Purity: ≥98%, CAS: 4836-13-9, 5 g
SDP
Catalog No. 502942014
Encompass_Preferred
A building block; has been used in the synthesis of nucleosides and nucleotides with anticancer activity, as well as oligonucleotides
Catalog No. 50-294-2014 Supplier Cayman Chemical Supplier No. 399345 G
A building block; has been used in the synthesis of nucleosides and nucleotides with anticancer activity, as well as oligonucleotides