Learn More
Medchemexpress LLC N4-Benzoyl-2 -O-meth 500mg
SDP
Catalog No. 5000204980
Encompass_Preferred
N4-Benzoyl-2 -O-methylcytidine is a natural cytidine nucleoside analog Cytidine analogs have the mechanism of inhibiting DNA methyltransferase (such as Zebularine (HY-13420)) and have potential antimetabolite and antitumor activities[1][2]
Catalog No. 50-002-04980 Supplier Medchemexpress LLC Supplier No. HYW114787500MG
N4-Benzoyl-2 -O-methylcytidine is a natural cytidine nucleoside analog Cytidine analogs have the mechanism of inhibiting DNA methyltransferase (such as Zebularine (HY-13420)) and have potential antimetabolite and antitumor activities[1][2]