Learn More
Medchemexpress LLC N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine | 67219-55-0 | 98.9% | 633.69 | 5 G
SDP
Catalog No. 5000452346
Encompass_Preferred
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine, also known as 5'-O-DMT-N4-Bz-dC, is a compound that can be used for the synthesis of oligodeoxynucleotides containing a 3'-S-phosphorothiolate (3'-PS) linkage. It serves as a useful tool for probing enzyme-catalyzed cleavage processes in DNA.
  • Used for synthesis of oligodeoxynucleotides.
  • Useful tool for probing enzyme-catalyzed cleavage processes in DNA.
Catalog No. 50-004-52346 Supplier Medchemexpress LLC Supplier No. HYW0107065G
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine, also known as 5'-O-DMT-N4-Bz-dC, is a compound that can be used for the synthesis of oligodeoxynucleotides containing a 3'-S-phosphorothiolate (3'-PS) linkage. It serves as a useful tool for probing enzyme-catalyzed cleavage processes in DNA.
  • Used for synthesis of oligodeoxynucleotides.
  • Useful tool for probing enzyme-catalyzed cleavage processes in DNA.