Learn More
Cayman Chemical N4-benzoyl-Cytidine, Purity: ≥98%, CAS: 13089-48-0, 250 g
SDP
Catalog No. 502910214
Encompass_Preferred
A benzyolated derivative of cytidine; a substrate of UCK1 and UCK2; has been used in the synthesis of 2ʹ-OH protecting groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV light-induced and targeted gene silencing in zebrafish embryos
Catalog No. 50-291-0214 Supplier Cayman Chemical Supplier No. 35698250 G
A benzyolated derivative of cytidine; a substrate of UCK1 and UCK2; has been used in the synthesis of 2ʹ-OH protecting groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV light-induced and targeted gene silencing in zebrafish embryos