Learn More
Cayman Chemical N4-benzoyl-CytIdIn 25g
SDP
Catalog No. 502911390
Encompass_Preferred
A benzyolated derivative of cytidine; a substrate of UCK1 and UCK2; has been used in the synthesis of 2ʹ-OH protecting groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV light-induced and targeted gene silencing in zebrafish embryos
Catalog No. 50-291-1390 Supplier Cayman Chemical Supplier No. 3569825 G
A benzyolated derivative of cytidine; a substrate of UCK1 and UCK2; has been used in the synthesis of 2ʹ-OH protecting groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV light-induced and targeted gene silencing in zebrafish embryos