Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. E065725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E065725G Supplier TCI America Supplier No. E065725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 25646-77-9
Melting Point 155°C
Color White-Yellow
Molecular Formula C11H20N2O5S
MDL Number MFCD00035563
Quantity 25 g
UN Number 2811
Synonym kodak cd 4, 4-n-ethyl-n-2-hydroxyethyl-2-methylphenylenediamine sulfate, n4-ethyl-n4-2-hydroxyethyl-2-methyl-1,4-phenylenediamine sulfate, cd 4, ppd 4 4, 2-4-amino-3-methylphenyl ethylamino ethanol sulfate, 2-4-amino-3-methylphenyl ethyl amino ethanol sulfate, 4-ammonio-m-tolyl ethyl 2-hydroxyethyl ammonium sulphate, n5-ethyl-n5-2'-hydroxyethyl toluene-2,5-diammonium sulfate
InChI Key GVEYRUKUJCHJSR-UHFFFAOYSA-N
SMILES OS(O)(=O)=O.CCN(CCO)C1=CC=C(N)C(C)=C1
IUPAC Name 2-[(4-amino-3-methylphenyl)(ethyl)amino]ethan-1-ol; sulfuric acid
Molecular Weight (g/mol) 292.35
PubChem CID 91579
Formula Weight 292.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine Sulfate
Show More Show Less
RTECSNumber KJ6477000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.