Learn More
Apexbio Technology LLC N4-Methyl-dCTP(Synonyms: N4-Methyl-2'-deoxycytidine-5'-triphosphate, N4-Me-dCTP, 4-Methyl-dCTP), 50ul (100 mM).
SDP
Catalog No. 5000060634
Encompass_Preferred
N4-Methyl-dCTP is a cytosine-derived modified deoxynucleoside triphosphate capable of being enzymatically incorporated into DNA strands thereby introducing cytosine methylation modifications at specific genomic sites DNA methylation at the N4 position of cytosine is observed naturally across both prokaryotic and eukaryotic organisms and influences processes such as epigenetic gene expression regulation DNA replication and DNA damage repair Researchers use N4-Methyl-dCTP to incorporate defined methylation modifications enabling investigation into methylation-specific DNA-binding protein interactions DNA methylation patterning analyses and functional studies of epigenetic modification pathways
Catalog No. 50-000-60634 Supplier Apexbio Technology LLC Supplier No. B81100.05
N4-Methyl-dCTP is a cytosine-derived modified deoxynucleoside triphosphate capable of being enzymatically incorporated into DNA strands thereby introducing cytosine methylation modifications at specific genomic sites DNA methylation at the N4 position of cytosine is observed naturally across both prokaryotic and eukaryotic organisms and influences processes such as epigenetic gene expression regulation DNA replication and DNA damage repair Researchers use N4-Methyl-dCTP to incorporate defined methylation modifications enabling investigation into methylation-specific DNA-binding protein interactions DNA methylation patterning analyses and functional studies of epigenetic modification pathways