Learn More
Cayman Chemical N4-Methylcytidine, Purity: ≥98%, CAS: 10578-79-7, 100 mg
SDP
Catalog No. 502945873
Encompass_Preferred
A pyrimidine nucleoside and derivative of cytidine; found in mammalian mitochondrial rRNA where it stabilizes the structure of 12S rRNA and is essential for mitochondrial translation of oxidative phosphorylation-related proteins; maintains normal base pairing with guanine in RNA oligomers
Catalog No. 50-294-5873 Supplier Cayman Chemical Supplier No. 40907100 MG
A pyrimidine nucleoside and derivative of cytidine; found in mammalian mitochondrial rRNA where it stabilizes the structure of 12S rRNA and is essential for mitochondrial translation of oxidative phosphorylation-related proteins; maintains normal base pairing with guanine in RNA oligomers