Learn More
Cayman Chemical N4-MethylcytIdIn 50mg
SDP
Catalog No. 502943168
Encompass_Preferred
A pyrimidine nucleoside and derivative of cytidine; found in mammalian mitochondrial rRNA where it stabilizes the structure of 12S rRNA and is essential for mitochondrial translation of oxidative phosphorylation-related proteins; maintains normal base pairing with guanine in RNA oligomers
Catalog No. 50-294-3168 Supplier Cayman Chemical Supplier No. 4090750 MG
A pyrimidine nucleoside and derivative of cytidine; found in mammalian mitochondrial rRNA where it stabilizes the structure of 12S rRNA and is essential for mitochondrial translation of oxidative phosphorylation-related proteins; maintains normal base pairing with guanine in RNA oligomers