Learn More
Medchemexpress LLC N4-methylcytidine | 10578-79-7 | MFCD23701766 | 100.0% | 257.24 | C10H15N3O5 | 1 ML
SDP
Catalog No. 5000283499
Encompass_Preferred
N4-methylcytidine is a modified cytidine nucleoside provided as a ready-to-use 10 mM solution in DMSO for research applications, commonly used in studies of RNA modification and nucleoside-related biochemical assays.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Useful for RNA modification and nucleoside biology studies.
  • SDS and COA available for download.
  • Multiple solid and solution pack sizes offered for scalability.
Catalog No. 50-002-83499 Supplier Medchemexpress LLC Supplier No. HYW13046610MM/1ML
Add to Cart
Add to Cart
N4-methylcytidine is a modified cytidine nucleoside provided as a ready-to-use 10 mM solution in DMSO for research applications, commonly used in studies of RNA modification and nucleoside-related biochemical assays.
  • Ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Useful for RNA modification and nucleoside biology studies.
  • SDS and COA available for download.
  • Multiple solid and solution pack sizes offered for scalability.