Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N4-methylcytidine | 10578-79-7 | MFCD23701766 | 100.0% | 257.24 g/mol | C10H15N3O5 | 250 MG
SDP

Catalog No. 5000283411
Encompass_Preferred

N4-methylcytidine is a cytidine nucleoside analog used as a research reagent to study nucleoside metabolism and DNA methylation-related processes. It is supplied as a high-purity solid with documented storage and solubility properties for use in biochemical and cellular assays.

  • Cytidine nucleoside analog for research use.
  • High purity (99.99%).
  • White to off-white solid appearance.
  • Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
  • Available in multiple package sizes, including 250 mg.

Catalog No. 50-002-83411 Supplier Medchemexpress LLC Supplier No. HYW130466250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N4-methylcytidine is a cytidine nucleoside analog used as a research reagent to study nucleoside metabolism and DNA methylation-related processes. It is supplied as a high-purity solid with documented storage and solubility properties for use in biochemical and cellular assays.

  • Cytidine nucleoside analog for research use.
  • High purity (99.99%).
  • White to off-white solid appearance.
  • Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
  • Available in multiple package sizes, including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.