Learn More
Cayman Chemical N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine, Purity: ≥98%, CAS: 67219-55-0, 100 mg
SDP
Catalog No. 502989648
Encompass_Preferred
A building block; has been used in the synthesis of oligodeoxyribonucleotides
Catalog No. 50-298-9648 Supplier Cayman Chemical Supplier No. 30803100 MG
A building block; has been used in the synthesis of oligodeoxyribonucleotides