Learn More
Cayman Chemical N-4-Tosyl-L-arginine methyl ester (hydrochloride), Purity: ≥98%, CAS: 1784-03-8, 50 g
SDP
Catalog No. 502967173
Encompass_Preferred
A synthetic substrate of serine proteases, including thrombin, trypsin, chymotrypsin, and kallikreins; also binds to and inhibits anaphase-promoting complex, a ubiquitin ligase, in Xenopus egg extract
Catalog No. 50-296-7173 Supplier Cayman Chemical Supplier No. 1755050 G
A synthetic substrate of serine proteases, including thrombin, trypsin, chymotrypsin, and kallikreins; also binds to and inhibits anaphase-promoting complex, a ubiquitin ligase, in Xenopus egg extract