Learn More
Cayman Chemical N4TosylLargInIn methyl e 5g
SDP
Supplier:  Cayman Chemical 175505 G
Encompass_Preferred
A synthetic substrate of serine proteases, including thrombin, trypsin, chymotrypsin, and kallikreins; also binds to and inhibits anaphase-promoting complex, a ubiquitin ligase, in Xenopus egg extract
Catalog No. 50-296-6253