Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N5-(1-imino-3-butenyl)-L-ornithine, monohydrochloride | 728944-69-2 | MFCD03410278 | 95.0% | 235.7 g/mol | C9H17N3O2·HCl | 5 MG
SDP

Catalog No. 5000262218
Encompass_Preferred

Vinyl-L-NIO hydrochloride is the monohydrochloride salt of Vinyl-L-NIO, a potent and selective neuronal nitric oxide synthase (nNOS) inhibitor used in biochemical and pharmacological research to reduce nitric oxide production. It is intended for research use in enzymatic assays, cell-based studies, and related laboratory applications; handle and store per supplier guidance.

  • Potent, selective nNOS inhibitor suitable for biochemical assays.
  • Reduces nitric oxide production in cellular and enzymatic studies.
  • High purity (≥95.0%) for reliable experimental results.
  • Molecular formula C9H17N3O2·HCl and molecular weight 235.7 g/mol.
  • Appearance reported as oil (off-white to light yellow).
  • Available in small research pack sizes such as 5 mg, 10 mg, and 50 mg.

Catalog No. 50-002-62218 Supplier Medchemexpress LLC Supplier No. HY1305795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vinyl-L-NIO hydrochloride is the monohydrochloride salt of Vinyl-L-NIO, a potent and selective neuronal nitric oxide synthase (nNOS) inhibitor used in biochemical and pharmacological research to reduce nitric oxide production. It is intended for research use in enzymatic assays, cell-based studies, and related laboratory applications; handle and store per supplier guidance.

  • Potent, selective nNOS inhibitor suitable for biochemical assays.
  • Reduces nitric oxide production in cellular and enzymatic studies.
  • High purity (≥95.0%) for reliable experimental results.
  • Molecular formula C9H17N3O2·HCl and molecular weight 235.7 g/mol.
  • Appearance reported as oil (off-white to light yellow).
  • Available in small research pack sizes such as 5 mg, 10 mg, and 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.