Learn More
Cayman Chemical N6- 2-amInoethyl-NAD 3.5mg
SDP
Catalog No. 502919120
Encompass_Preferred
A derivative of NAD+; has been conjugated to various matrices, including sepharose and dextran, for affinity chromatography of NAD+-dependent enzymes; has been conjugated to silylated chips via PQQ to act as a biosensor for lactate
Catalog No. 50-291-9120 Supplier Cayman Chemical Supplier No. 385693.5 MG
A derivative of NAD+; has been conjugated to various matrices, including sepharose and dextran, for affinity chromatography of NAD+-dependent enzymes; has been conjugated to silylated chips via PQQ to act as a biosensor for lactate