Learn More
Medchemexpress LLC N6-(2-Hydroxyethyl)adenosine | 4338-48-1 | 311.30 | 100 MG
SDP
Catalog No. 5000460374
Encompass_Preferred
N6-(2-Hydroxyethyl)adenosine is a purine nucleoside analog that inhibits the NF-κB/Smad signaling pathway. It demonstrates various biological activities, including anti-hyperglycemia, antioxidant, antitumor, anti-inflammatory, and insecticidal effects, and is orally active.
  • Reduces LPS-induced TNF-α and IL-1β expression
  • Increases IL-10 secretion in RAW 264.7 cells
  • Reduces TGF-β1-induced expression of collagen I, α-SMA, and fibronectin in NRK-49F cells
  • Induces ROS generation and decreases mitochondrial membrane potential
  • Exhibits cytotoxicity in SGC-7901 and AGS cancer cells
  • Leads to apoptosis and autophagy
  • Exhibits insecticidal activity against *Plutella xylostella*
  • Reduces blood glucose, serum creatinine, blood urea nitrogen, and urine protein levels in diabetic rats
  • Alleviates glomeruli and renal tubule damage
  • Reduces tumor volume and weight in SGC-7901 xenograft mouse models
Catalog No. 50-004-60374 Supplier Medchemexpress LLC Supplier No. HYW006957100MG
N6-(2-Hydroxyethyl)adenosine is a purine nucleoside analog that inhibits the NF-κB/Smad signaling pathway. It demonstrates various biological activities, including anti-hyperglycemia, antioxidant, antitumor, anti-inflammatory, and insecticidal effects, and is orally active.
  • Reduces LPS-induced TNF-α and IL-1β expression
  • Increases IL-10 secretion in RAW 264.7 cells
  • Reduces TGF-β1-induced expression of collagen I, α-SMA, and fibronectin in NRK-49F cells
  • Induces ROS generation and decreases mitochondrial membrane potential
  • Exhibits cytotoxicity in SGC-7901 and AGS cancer cells
  • Leads to apoptosis and autophagy
  • Exhibits insecticidal activity against *Plutella xylostella*
  • Reduces blood glucose, serum creatinine, blood urea nitrogen, and urine protein levels in diabetic rats
  • Alleviates glomeruli and renal tubule damage
  • Reduces tumor volume and weight in SGC-7901 xenograft mouse models