Learn More
Cayman Chemical N6- 2-IsopentenyladenIn 1g
SDP
Catalog No. 502967929
Encompass_Preferred
A naturally occurring cytokinin that regulates cell division, development, and nutrient processing in plants and serves as a precursor to zeatin synthesis
Catalog No. 50-296-7929 Supplier Cayman Chemical Supplier No. 179061 G
A naturally occurring cytokinin that regulates cell division, development, and nutrient processing in plants and serves as a precursor to zeatin synthesis