Learn More
Cayman Chemical N6- 2-IsopentenyladenIn 5g
SDP
Catalog No. 502965320
Encompass_Preferred
A naturally occurring cytokinin that regulates cell division, development, and nutrient processing in plants and serves as a precursor to zeatin synthesis
Catalog No. 50-296-5320 Supplier Cayman Chemical Supplier No. 179065 G
A naturally occurring cytokinin that regulates cell division, development, and nutrient processing in plants and serves as a precursor to zeatin synthesis