Learn More
Cayman Chemical N6- 2-IsopentenyladenosIn 50mg
SDP
Catalog No. 502984867
Encompass_Preferred
A precursor in the biosynthesis of the plant hormone N6-(Δ2-isopentenyl)adenine, which regulates diverse events in plant growth and development; can also alter post-transcriptional processes in mammalian cells
Catalog No. 50-298-4867 Supplier Cayman Chemical Supplier No. 2052250 MG
A precursor in the biosynthesis of the plant hormone N6-(Δ2-isopentenyl)adenine, which regulates diverse events in plant growth and development; can also alter post-transcriptional processes in mammalian cells