Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6,2'-O-Dibutyryladenosine 3',5'-Cyclic Monophosphate Sodium Salt 97.0+%, TCI America™
SDP

Catalog No. D422825MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D422825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D422825MG Supplier TCI America Supplier No. D422825MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16980-89-5
Molecular Formula C18H26N5NaO8P
Molecular Weight (g/mol) 494.397
MDL Number MFCD00005843
InChI Key KSSDVZRQZQECKZ-VMZSDVDQSA-N
Synonym bucladesine
PubChem CID 131673831
IUPAC Name [(4aR,6R,7R,7aR)-6-[6-(butanoylamino)purin-9-yl]-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-yl] butanoate;molecular hydrogen;sodium
SMILES [HH].CCCC(=O)NC1=NC=NC2=C1N=CN2C3C(C4C(O3)COP(=O)(O4)O)OC(=O)CCC.[Na]

Specifications

Color White
Quantity 25 mg
Formula Weight 491.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N6,2′-O-Dibutyryladenosine 3′,5′-Cyclic Monophosphate Sodium Salt
RTECSNumber ES5055500
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.