Learn More
Sigma Aldrich Fine Chemicals Biosciences N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, 16980-89-5, MFCD00005843, 250mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D0627250MG
Encompass
Molecular Formula: C18H23N5O8PNa, Molecular Weight: 491.37, Assay: ≥96% (HPLC), Solubility: H2O: 50 mg/mL
Catalog No. NC0338844