Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-(2-phenylethyl)adenosine | 20125-39-7 | MFCD00055128 | 99.4% | 371.39 g/mol | C18H21N5O4 | 200 MG
SDP

Catalog No. 5000248137
Encompass_Preferred

N6-(2-Phenylethyl)adenosine is an adenosine derivative provided as a research-grade small molecule for studies of adenosine receptor pharmacology. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research use.

  • CAS number 20125-39-7.
  • Molecular formula C18H21N5O4.
  • Molecular weight 371.39 g/mol.
  • Purity 99.4%.
  • Physical form white to off-white solid.
  • Pack size 200 mg.
  • Storage powder: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-48137 Supplier Medchemexpress LLC Supplier No. HY101854200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-(2-Phenylethyl)adenosine is an adenosine derivative provided as a research-grade small molecule for studies of adenosine receptor pharmacology. It is supplied as a white to off-white solid with high reported purity and is intended for laboratory research use.

  • CAS number 20125-39-7.
  • Molecular formula C18H21N5O4.
  • Molecular weight 371.39 g/mol.
  • Purity 99.4%.
  • Physical form white to off-white solid.
  • Pack size 200 mg.
  • Storage powder: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.