Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-[(2E)-1-oxo-2-buten-1-yl]-L-lysine | 1338823-35-0 | MFCD22381072 | 99.4% | 214.26 g/mol | C10H18N2O3 | 1 G
SDP

Catalog No. 5000210335
Encompass_Preferred

N6-(But-2-enoyl)-L-lysine is a chemically modified lysine derivative supplied as a solid powder for research use. It has a defined molecular formula, molecular weight, and high purity, with recommended storage conditions to maintain stability.

  • CAS number: 1338823-35-0.
  • Molecular formula: C10H18N2O3.
  • Molecular weight: 214.26 g/mol.
  • Purity: 99.43%.
  • Available sizes: 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.
  • Appearance: white to off-white solid powder.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-10335 Supplier Medchemexpress LLC Supplier No. HYW0482031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N6-(But-2-enoyl)-L-lysine is a chemically modified lysine derivative supplied as a solid powder for research use. It has a defined molecular formula, molecular weight, and high purity, with recommended storage conditions to maintain stability.

  • CAS number: 1338823-35-0.
  • Molecular formula: C10H18N2O3.
  • Molecular weight: 214.26 g/mol.
  • Purity: 99.43%.
  • Available sizes: 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g.
  • Appearance: white to off-white solid powder.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.