Learn More
Cayman Chemical N6-(Δ2-Isopentenyl)adenosine, Purity: ≥98%, CAS: 7724-76-7, 250 mg
SDP
Catalog No. 502984716
Encompass_Preferred
A precursor in the biosynthesis of the plant hormone N6-(Δ2-isopentenyl)adenine, which regulates diverse events in plant growth and development; can also alter post-transcriptional processes in mammalian cells
Catalog No. 50-298-4716 Supplier Cayman Chemical Supplier No. 20522250 MG
A precursor in the biosynthesis of the plant hormone N6-(Δ2-isopentenyl)adenine, which regulates diverse events in plant growth and development; can also alter post-transcriptional processes in mammalian cells