Learn More
Medchemexpress LLC N6-(4-Hydroxybenzyl)adenosine | 110505-75-4 | 99.96% | 373.36 | 100 MG
SDP
Catalog No. 5000387921
Encompass_Preferred
N6-(4-Hydroxybenzyl)adenosine inhibits platelet aggregation induced by collagen, potentially by interacting with the P2Y12 receptor binding site. It also prevents serum deprivation-induced apoptosis in PC-12 cells.
  • Inhibits platelet aggregation
  • Prevents serum deprivation-induced apoptosis in PC-12 cells
  • Active against collagen-induced aggregation
Catalog No. 50-003-87921 Supplier Medchemexpress LLC Supplier No. HY18775100MG
N6-(4-Hydroxybenzyl)adenosine inhibits platelet aggregation induced by collagen, potentially by interacting with the P2Y12 receptor binding site. It also prevents serum deprivation-induced apoptosis in PC-12 cells.
  • Inhibits platelet aggregation
  • Prevents serum deprivation-induced apoptosis in PC-12 cells
  • Active against collagen-induced aggregation