Learn More
Cayman Chemical N6-(4-Aminobutyl)adenosine-5'-O-triphosphate (sodium salt), Purity: ≥95%, 2.9 mg
SDP
Catalog No. 502919080
Encompass_Preferred
A derivative of ATP; contains an N6 butyl substitution with a terminal amino moiety
Catalog No. 50-291-9080 Supplier Cayman Chemical Supplier No. 385412.9 MG
A derivative of ATP; contains an N6 butyl substitution with a terminal amino moiety