Learn More
Cayman Chemical N6-[(6-Aminohexyl)carbamoylmethyl]-ADP, Purity: ≥70%, CAS: 100826-95-7, 5 mg
SDP
Catalog No. 502918723
Encompass_Preferred
An alkylated ADP derivative; has been biotinylated and conjugated to an azidonitrobenzoyl group for use as a photoaffinity probe to visualize the ATPase site of rabbit myosin
Catalog No. 50-291-8723 Supplier Cayman Chemical Supplier No. 384025 MG
An alkylated ADP derivative; has been biotinylated and conjugated to an azidonitrobenzoyl group for use as a photoaffinity probe to visualize the ATPase site of rabbit myosin