Learn More
Cayman Chemical N6-benzoyl-2'-Deoxyadenosine, Purity: ≥95%, CAS: 4546-72-9, 1 g
SDP
Catalog No. 502988183
Encompass_Preferred
A nucleoside building block; has been used in the synthesis of various oligonucleotides
Catalog No. 50-298-8183 Supplier Cayman Chemical Supplier No. 302621 G
A nucleoside building block; has been used in the synthesis of various oligonucleotides