Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6-Benzoyl-2'-deoxyadenosine, 98%, Thermo Scientific™

Catalog No. AAJ6417506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6417506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6417506 Supplier Thermo Scientific Chemicals Supplier No. J6417506
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N6-Benzoyl-2′-deoxyadenosine is used in formation of N-Branched oligonucleotides as by-products in solid-phase oligonucleotide synthesis.

Solubility
Soluble in water (slightly), and methanol (20 mg/ml).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4546-72-9
Molecular Formula C17H17N5O4
Molecular Weight (g/mol) 355.35
MDL Number MFCD00009628
InChI Key PIXHJAPVPCVZSV-LJEKBPCBNA-N
Synonym n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine
PubChem CID 107558
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

Color White
Quantity 5 g
Formula Weight 355.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N6-Benzoyl-2'-deoxyadenosine
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.