Learn More
Medchemexpress LLC N6-benzoyl-2'-deoxyadenosine monohydrate | 305808-19-9 | 98.0% | 373.36 | C17H19N5O5 | 25 G
SDP
Catalog No. 5000283621
Encompass_Preferred
N6-benzoyl-2'-deoxyadenosine monohydrate is a nucleoside analog used in biochemical research and oligonucleotide synthesis. Supplied as a solid monohydrate, it is used in diagnostic and analytical workflows that require modified nucleosides.
  • Nucleoside analog used in oligonucleotide synthesis and nucleoside research.
  • Monohydrate solid form for easier handling and storage.
  • High purity (98.0%) suitable for research applications.
  • Chemical formula C17H19N5O5; molecular weight 373.36.
  • Available in common laboratory pack sizes, including 25 G.
  • Applicable in diagnostic and analytical workflows requiring DNA binding or modification.
Catalog No. 50-002-83621 Supplier Medchemexpress LLC Supplier No. HYW14089725G
N6-benzoyl-2'-deoxyadenosine monohydrate is a nucleoside analog used in biochemical research and oligonucleotide synthesis. Supplied as a solid monohydrate, it is used in diagnostic and analytical workflows that require modified nucleosides.
  • Nucleoside analog used in oligonucleotide synthesis and nucleoside research.
  • Monohydrate solid form for easier handling and storage.
  • High purity (98.0%) suitable for research applications.
  • Chemical formula C17H19N5O5; molecular weight 373.36.
  • Available in common laboratory pack sizes, including 25 G.
  • Applicable in diagnostic and analytical workflows requiring DNA binding or modification.