Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-benzoyl-2'-deoxyadenosine monohydrate | 305808-19-9 | MFCD00009628 | ≥98.0% | 373.37 g/mol | C17H19N5O5 | 1 G
SDP

Catalog No. 5000282804
Encompass_Preferred

N6-Benzoyl-2'-deoxyadenosine monohydrate is a purine nucleoside analog provided as a monohydrate solid for research and diagnostic applications. It is used to study DNA duplex interactions and related biochemical assays, and is supplied at research-grade purity for laboratory use.

  • High purity (≥98.0%).
  • Monohydrate solid form suitable for laboratory assays.
  • Used as a nucleoside analog to probe DNA duplex structure in assays.
  • CAS number 305808-19-9.
  • Molecular weight 373.37 g/mol.
  • Available in a 1 g pack and larger sizes.

Catalog No. 50-002-82804 Supplier Medchemexpress LLC Supplier No. HYW1408971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-Benzoyl-2'-deoxyadenosine monohydrate is a purine nucleoside analog provided as a monohydrate solid for research and diagnostic applications. It is used to study DNA duplex interactions and related biochemical assays, and is supplied at research-grade purity for laboratory use.

  • High purity (≥98.0%).
  • Monohydrate solid form suitable for laboratory assays.
  • Used as a nucleoside analog to probe DNA duplex structure in assays.
  • CAS number 305808-19-9.
  • Molecular weight 373.37 g/mol.
  • Available in a 1 g pack and larger sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.