Learn More
Medchemexpress LLC N6-benzoyl-2'-deoxyadenosine monohydrate | 305808-19-9 | MFCD00149575 | 98.0% | 373.36 g/mol | C17H19N5O5 | 10 G
SDP
Catalog No. 5000282871
Encompass_Preferred
N6-benzoyl-2'-deoxyadenosine monohydrate is a protected nucleoside analog supplied as a monohydrate solid for research use. It serves as a building block in oligonucleotide and nucleotide chemistry and is characterized for purity and composition to support analytical and synthetic workflows.
  • Protected adenosine derivative for nucleotide synthesis.
  • Monohydrate solid, white to off-white in appearance.
  • High purity (98.0%) for reproducible results.
  • Available in multiple package sizes, including 10 G, for scale flexibility.
  • Documented molecular weight and formula for formulation and analysis.
Catalog No. 50-002-82871 Supplier Medchemexpress LLC Supplier No. HYW14089710G
N6-benzoyl-2'-deoxyadenosine monohydrate is a protected nucleoside analog supplied as a monohydrate solid for research use. It serves as a building block in oligonucleotide and nucleotide chemistry and is characterized for purity and composition to support analytical and synthetic workflows.
  • Protected adenosine derivative for nucleotide synthesis.
  • Monohydrate solid, white to off-white in appearance.
  • High purity (98.0%) for reproducible results.
  • Available in multiple package sizes, including 10 G, for scale flexibility.
  • Documented molecular weight and formula for formulation and analysis.