Learn More
Medchemexpress LLC N6-benzoyl-2'-deoxyadenosine monohydrate | 305808-19-9 | MFCD00149575 | >98.0% | 373.4 g/mol | C17H19N5O5 | 5 G
SDP
Catalog No. 5000283622
Encompass_Preferred
N6-Benzoyl-2'-deoxyadenosine monohydrate is a nucleoside analog of deoxyadenosine bearing an N6-benzoyl protecting group. It is provided as a monohydrate solid for research use (CAS 305808-19-9) with purity ≥98.0%, and is used as an intermediate in nucleoside chemistry, oligonucleotide synthesis, and studies of nucleoside analog activity.
  • High purity suitable for research applications.
  • Monohydrate solid form for stable handling and storage.
  • Serves as an intermediate in nucleoside and oligonucleotide synthesis.
  • Compatible with standard analytical and synthetic workflows.
  • Supplied in small-scale laboratory pack sizes for bench use.
Catalog No. 50-002-83622 Supplier Medchemexpress LLC Supplier No. HYW1408975G
N6-Benzoyl-2'-deoxyadenosine monohydrate is a nucleoside analog of deoxyadenosine bearing an N6-benzoyl protecting group. It is provided as a monohydrate solid for research use (CAS 305808-19-9) with purity ≥98.0%, and is used as an intermediate in nucleoside chemistry, oligonucleotide synthesis, and studies of nucleoside analog activity.
  • High purity suitable for research applications.
  • Monohydrate solid form for stable handling and storage.
  • Serves as an intermediate in nucleoside and oligonucleotide synthesis.
  • Compatible with standard analytical and synthetic workflows.
  • Supplied in small-scale laboratory pack sizes for bench use.