Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-benzoyl-2'-fluoro-2'-deoxyadenosine | 136834-20-3 | MFCD22373350 | 99.6% | 373.34 g·mol^-1 | C17H16FN5O4 | 250 MG
SDP

Catalog No. 5000209686
Encompass_Preferred

2'-F-Bz-A (N6-benzoyl-2'-fluoro-2'-deoxyadenosine) is a nucleoside analog used as a building block in oligoribonucleotide synthesis and nucleic-acid chemistry. It is supplied as a solid reagent with high chemical purity for research applications.

  • Nucleoside analog suitable for oligoribonucleotide synthesis.
  • High purity (99.6%) for reliable experimental results.
  • Molecular formula C17H16FN5O4; molecular weight 373.34 g·mol^-1.
  • Commonly used as a reagent in nucleic-acid chemistry and oligonucleotide modification.
  • Available in multiple quantities appropriate for laboratory research.

Catalog No. 50-002-09686 Supplier Medchemexpress LLC Supplier No. 250MGHYW089288
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-F-Bz-A (N6-benzoyl-2'-fluoro-2'-deoxyadenosine) is a nucleoside analog used as a building block in oligoribonucleotide synthesis and nucleic-acid chemistry. It is supplied as a solid reagent with high chemical purity for research applications.

  • Nucleoside analog suitable for oligoribonucleotide synthesis.
  • High purity (99.6%) for reliable experimental results.
  • Molecular formula C17H16FN5O4; molecular weight 373.34 g·mol^-1.
  • Commonly used as a reagent in nucleic-acid chemistry and oligonucleotide modification.
  • Available in multiple quantities appropriate for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.