Learn More
Medchemexpress LLC N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine | 64325-78-6 | 98.0% | 657.71 | 1 ML
SDP
Catalog No. 5000454615
Encompass_Preferred
N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine serves as an intermediate in various chemical processes. It is intended for research use only and is not sold to patients.
  • Used as a chemical intermediate.
  • High purity of 98.0%.
  • Available in different forms including solid and a 10 mM solution in DMSO.
  • Detailed solubility information and stock solution preparation guidance provided.
  • Comprehensive documentation including Data Sheet, COA, SDS, and Handling Instructions.
Catalog No. 50-004-54615 Supplier Medchemexpress LLC Supplier No. HYW01307710MM/1ML
N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine serves as an intermediate in various chemical processes. It is intended for research use only and is not sold to patients.
  • Used as a chemical intermediate.
  • High purity of 98.0%.
  • Available in different forms including solid and a 10 mM solution in DMSO.
  • Detailed solubility information and stock solution preparation guidance provided.
  • Comprehensive documentation including Data Sheet, COA, SDS, and Handling Instructions.