Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine, 98%

Catalog No. AAJ6509803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6509803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6509803 Supplier Thermo Scientific Chemicals Supplier No. J6509803
Add to Cart

C39H37N5O7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine
Color White to Yellow
Molecular Formula C39H37N5O7
Quantity 1 g
MDL Number MFCD08704196
Solubility Information Soluble in chloroform at 50mg/ml
Formula Weight 687.74
Percent Purity 98%
Physical Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.