Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite | 179479-02-8 | 98.0% | C48H54N7O8P | 25 MG
SDP

Catalog No. 5000358953
Encompass_Preferred

N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite is an adenosine analog that functions as a smooth muscle vasodilator and has demonstrated potential in inhibiting cancer progression. It is a white to off-white solid with a molecular weight of 887.96 and a purity of 98.04%. This product is intended for research use only and is not for sale to patients.

  • Molecular weight: 887.96
  • Formula: C48H54N7O8P
  • CAS No.: 179479-02-8
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.04%
  • Biological activity: Acts as a smooth muscle vasodilator and inhibits cancer progression.
  • Solubility: Soluble in DMSO at 100 mg/mL (112.62 mM) with ultrasonic assistance, recommending the use of newly opened DMSO due to the impact of hygroscopic DMSO on solubility.

Catalog No. 50-003-58953 Supplier Medchemexpress LLC Supplier No. 25MGHY45281
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite is an adenosine analog that functions as a smooth muscle vasodilator and has demonstrated potential in inhibiting cancer progression. It is a white to off-white solid with a molecular weight of 887.96 and a purity of 98.04%. This product is intended for research use only and is not for sale to patients.

  • Molecular weight: 887.96
  • Formula: C48H54N7O8P
  • CAS No.: 179479-02-8
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.04%
  • Biological activity: Acts as a smooth muscle vasodilator and inhibits cancer progression.
  • Solubility: Soluble in DMSO at 100 mg/mL (112.62 mM) with ultrasonic assistance, recommending the use of newly opened DMSO due to the impact of hygroscopic DMSO on solubility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.