Learn More
Medchemexpress LLC N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite | 179479-02-8 | 98.04% | C48H54N7O8P | 100 MG
SDP
Catalog No. 5000359060
Encompass_Preferred
N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite is an adenosine analog that primarily acts as a smooth muscle vasodilator. It has also been shown to inhibit cancer progression. This product is for research use only.
  • Appears as a solid with a color ranging from white to off-white
  • Molecular weight: 887.96
  • Primarily functions as a smooth muscle vasodilator
  • Demonstrated ability to inhibit cancer progression
Catalog No. 50-003-59060 Supplier Medchemexpress LLC Supplier No. HY45281100MG
N6-Benzoyl-5'-O-DMT-3'-O-methyladenosine 3'CE-phosphoramidite is an adenosine analog that primarily acts as a smooth muscle vasodilator. It has also been shown to inhibit cancer progression. This product is for research use only.
  • Appears as a solid with a color ranging from white to off-white
  • Molecular weight: 887.96
  • Primarily functions as a smooth muscle vasodilator
  • Demonstrated ability to inhibit cancer progression