Learn More
Medchemexpress LLC N6-Benzoyl-9-β-D-arabinofuranosyladenine | 79896-97-2 | 98.2% | C17H17N5O5 | 25 MG
SDP
Catalog No. 5000358555
Encompass_Preferred
N6-Benzoyl-9-β-D-arabinofuranosyladenine is an adenosine analog that primarily targets adenosine receptors. This compound exhibits smooth muscle vasodilatory effects and has shown potential in inhibiting cancer progression. It serves as a valuable tool for research in cardiovascular and oncology fields, enabling exploration of adenosine signaling pathways.
  • Adenosine analog
  • Primarily targets adenosine receptors
  • Exhibits smooth muscle vasodilatory effects
  • Demonstrates potential in inhibiting cancer progression
  • Valuable tool for cardiovascular research
  • Valuable tool for oncology research
  • Enables exploration of therapeutic implications of adenosine signaling pathways
Catalog No. 50-003-58555 Supplier Medchemexpress LLC Supplier No. 25MGHY47272
N6-Benzoyl-9-β-D-arabinofuranosyladenine is an adenosine analog that primarily targets adenosine receptors. This compound exhibits smooth muscle vasodilatory effects and has shown potential in inhibiting cancer progression. It serves as a valuable tool for research in cardiovascular and oncology fields, enabling exploration of adenosine signaling pathways.
  • Adenosine analog
  • Primarily targets adenosine receptors
  • Exhibits smooth muscle vasodilatory effects
  • Demonstrates potential in inhibiting cancer progression
  • Valuable tool for cardiovascular research
  • Valuable tool for oncology research
  • Enables exploration of therapeutic implications of adenosine signaling pathways