Learn More
Cayman Chemical N6-Benzyladenine, Purity: ≥98%, CAS: 1214-39-7, 100 mg
SDP
Catalog No. 502986536
Encompass_Preferred
A plant growth regulator; it deepens color and increases firmness, as well as decreases fibrous hardness, in asparagus at 20 ppm postharvest; induces shoot multiplication and shoot growth from turmeric and ginger seeds at 1 and 2 mg/L; increases wart density on cucumbers at 100 µM; decreases proliferation of HL-60 cells (IC50 = 67.6 µM)
Catalog No. 50-298-6536 Supplier Cayman Chemical Supplier No. 21711100 MG
A plant growth regulator; it deepens color and increases firmness, as well as decreases fibrous hardness, in asparagus at 20 ppm postharvest; induces shoot multiplication and shoot growth from turmeric and ginger seeds at 1 and 2 mg/L; increases wart density on cucumbers at 100 µM; decreases proliferation of HL-60 cells (IC50 = 67.6 µM)