Learn More
Medchemexpress LLC N6-(carboxymethyl)-L-lysine-d3 | 2699607-49-1 | 99.0% | 207.24 | C8H13D3N2O4 | 1 MG
SDP
Catalog No. 5000281945
Encompass_Preferred
CML-d3 is a deuterium-labeled stable isotope standard of carboxymethyl-lysine designed for use as a tracer or internal standard in analytical workflows such as mass spectrometry and quantitative biochemical assays. It is supplied as a powder with supporting documentation for analytical use and safe handling.
Catalog No. 50-002-81945 Supplier Medchemexpress LLC Supplier No. HYW286743S11MG
CML-d3 is a deuterium-labeled stable isotope standard of carboxymethyl-lysine designed for use as a tracer or internal standard in analytical workflows such as mass spectrometry and quantitative biochemical assays. It is supplied as a powder with supporting documentation for analytical use and safe handling.