Learn More
Cayman Chemical N6-Cyclopentyladenosine, Purity: ≥98%, CAS: 41552-82-3, 100 mg
SDP
Catalog No. 502993987
Encompass_Preferred
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)
Catalog No. 50-299-3987 Supplier Cayman Chemical Supplier No. 21448100 MG
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)