Learn More
Cayman Chemical N6-CyclopentyladenosIn 10mg
SDP
Catalog No. 502964517
Encompass_Preferred
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)
Catalog No. 50-296-4517 Supplier Cayman Chemical Supplier No. 2144810 MG
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)