Learn More
Cayman Chemical N6-CyclopentyladenosIn 50mg
SDP
Catalog No. 502964364
Encompass_Preferred
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)
Catalog No. 50-296-4364 Supplier Cayman Chemical Supplier No. 2144850 MG
A selective agonist at A1R (Kis = 2.3, 790, and 43 nM for A1R, A2R, and A3R receptors, respectively)