Learn More
Medchemexpress LLC N6-cyclopentyladenosine | 41552-82-3 | MFCD00036845 | 100.0% | 335.36 g·mol⁻1 | C15H21N5O4 | 100 MG
SDP
Catalog No. 5000247916
Encompass_Preferred
N6-Cyclopentyladenosine is a selective adenosine A1 receptor agonist used in pharmacology research and receptor signaling studies. It is supplied as a white to off-white solid with a molecular formula of C15H21N5O4 and a molecular weight of 335.36 g·mol⁻1.
  • Selective adenosine A1 receptor agonist with high affinity for A1 receptors.
  • Suitable for in vitro and in vivo receptor pharmacology studies.
  • White to off-white solid form convenient for handling and dosing.
  • Available in multiple sizes, including 100 MG packaging for preparative use.
  • Stable under recommended storage conditions for reliable experimental results.
Catalog No. 50-002-47916 Supplier Medchemexpress LLC Supplier No. HY103181100MG
N6-Cyclopentyladenosine is a selective adenosine A1 receptor agonist used in pharmacology research and receptor signaling studies. It is supplied as a white to off-white solid with a molecular formula of C15H21N5O4 and a molecular weight of 335.36 g·mol⁻1.
  • Selective adenosine A1 receptor agonist with high affinity for A1 receptors.
  • Suitable for in vitro and in vivo receptor pharmacology studies.
  • White to off-white solid form convenient for handling and dosing.
  • Available in multiple sizes, including 100 MG packaging for preparative use.
  • Stable under recommended storage conditions for reliable experimental results.