Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N6-cyclopentyladenosine | 41552-82-3 | MFCD00036845 | 100.0% | 335.36 g/mol | C15H21N5O4 | 1 ML
SDP

Catalog No. 5000247745
Encompass_Preferred

N6-Cyclopentyladenosine is a selective adenosine A1 receptor agonist used as a research reagent in pharmacology and receptor signaling studies. Supplied as a 10 mM solution in a 1 mL vial, it is intended for in vitro experiments and receptor-binding assays.

  • Selective adenosine A1 receptor agonist for pharmacological studies.
  • Provided as a 10 mM solution for ready-to-use experiments.
  • High chemical purity suitable for research applications.
  • Compact 1 mL vial format for small-scale assays.
  • Molecular weight 335.36 g/mol; molecular formula C15H21N5O4.

Catalog No. 50-002-47745 Supplier Medchemexpress LLC Supplier No. HY10318110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-Cyclopentyladenosine is a selective adenosine A1 receptor agonist used as a research reagent in pharmacology and receptor signaling studies. Supplied as a 10 mM solution in a 1 mL vial, it is intended for in vitro experiments and receptor-binding assays.

  • Selective adenosine A1 receptor agonist for pharmacological studies.
  • Provided as a 10 mM solution for ready-to-use experiments.
  • High chemical purity suitable for research applications.
  • Compact 1 mL vial format for small-scale assays.
  • Molecular weight 335.36 g/mol; molecular formula C15H21N5O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.